VWR® Maximum Protection Cut Protection Gloves
Supplier: VWR International
Certificates
About this item
The VWR® maximum cut protection glove with polyureathane coating provides superior cut protection for applications that include a high risk of cuts.
- Nylon/spandex/polyester blend composition
- Polyurethane (PU) coated
- Gray knit glove with gray coating
Ideal for use in environments handling items or tools with a high risk of cuts, including steel or glass handling, assembly, maintenance, and janitorial applications.
Certifications: Meets requirements for ANSI A5 Cut Protection (Level E). EN388:2016. The CE marking on these products means that they meet the requirements of European directive 89/686/CEE or EU Regulation 2016/425 on Personal Protective Equipment concerning protection, comfort and strength. Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Specifications
- Catalog No:
- 76408-554
- Latex/Latex-free:
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Finish:
- Polyurethane coating
- Gauge:
- 13
- Sterility:
- Non sterile