VWR® Cut Protection Gloves, NBR Coating
Fournisseur: VWR International
Certificats
À propos de cet article
The VWR® cut protection glove with nitrile coating provides general cut protection for the widest range of applications.
- Nylon/spandex/polyester blend composition
- Nitrile (NBR) coated
- Gray knit glove with black coating
Ideal for use in environments handling items or tools with a risk of cuts, including material handling, assembly, maintenance, and janitorial applications.
Certifications: Meets requirements for ANSI A2 Cut Protection (Level B). EN388:2016. The CE marking on these products means that they meet the requirements of European directive 89/686/CEE or EU Regulation 2016/425 on Personal Protective Equipment concerning protection, comfort and strength. Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.
Spécifications
- Catalog No:
- 76408-530
- 76408-528
- Latex/Latex-free:
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Finish:
- Nitrile coating
- Nitrile coating
- Gauge:
- 13
- 13
- Sterility:
- Non sterile
- Non sterile