Ward's OneRack | Avantor
Ward's OneRack
Supplier:   Heathrow Scientific
Ward's OneRack
Certificates
Ward's OneRack
2 Options Available Below
Product Details & Documents
The most flexible tube rack designed for use with a multitude of tube diameters and heights.
  • Interchangeable inserts accommodate multiple sized tubes
  • Inserts can be pivoted as needed
  • Open design allows visibility of tube graduations when filling
  • Teardrop shaped wells allow better retention of tubes
The Ward's® OneRack and its interchangeable shelves can accommodate virtually any brand tube (10 to 30 mm) from 5 to 50 ml volume, even if they have varying heights. Inserts can be pivoted in either an upper or lower position to securely accommodate the size and height of tube being used.
Product Family Options
Informations sur le produit
  • Désignation
    For Tubes
    L×W×H
    Availability
    Price
  • Ward's® OneRack (blue/clear)
    5 - 50 ml
    12.2×4.2×3"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 80,60 $
    80,60 $
    Log in pour la disponibilité
    Case of 50
    Case of 50 - 4 030,00 $
    4 030,00 $
    Each
       80,60 $
    Case of 50
       4 030,00 $
    Supplier Number: 120817W
  • Ward's® OneRack (purple/clear)
    5 - 50 ml
    12.2×4.2×3"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 80,60 $
    80,60 $
    Log in pour la disponibilité
    Case of 50
    Case of 50 - 4 030,00 $
    4 030,00 $
    Each
       80,60 $
    La quantité minimale de commande est de 1
    Case of 50
       4 030,00 $
    La quantité minimale de commande est de 1
    Supplier Number: 120818W