VWR® Plus Series Solid Door Undercounter Built-in ADA Compliant Refrigerators
Fournisseur: VWR International
Certificats
À propos de cet article
These undercounter refrigerators provide lab quality cold storage with ADA compliance.
- Integrated temperature alarms
- Keyed door locks
- Forced draft circulation
- Precise microprocessor temperature control
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
Built-in units are designed with front-facing ventilation systems, allowing easy installation into a bench or countertop. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 76422-930, 76422-932
Ordering information: All units have a two-year parts and labor warranty, plus an additional three year compressor parts warranty.
Spécifications
- Catalog No:
- 76422-932
- 76422-930
- Cat. No.:
- 76422-932
- 76422-930
- Defrost:
- Cycle
- Cycle
- Shelves/drawers:
- 2 adjustable, 1 fixed
- 2 adjustable, 1 fixed
- Door type:
- One solid swing door, left hinged
- One solid swing door, right hinged
- Electrical:
- 115 V, 60 Hz, 1.74 A, 1/8 HP
- 115 V, 60 Hz, 1.74 A, 1/8 HP
- Shipping weight:
- 68 kg (150 lbs.)
- 68 kg (150 lbs.)
- Environmental certification:
- HFC-free R600a refrigerant
- HFC-free R600a refrigerant