VWR® Maximum Protection Sterile Duckbill Mask with Ties
Catalogue # 76420-850
Fournisseur: VWR International
Spécifications
- Sterility:Sterile
- Color:White
- Description:Duckbill mask with ties
- Cat. no.:76420-850
Spécifications
À propos de cet article
The new sterile duckbill mask with ties expands upon the VWR® mask types in the popular cleanroom Maximum Protection line by providing a sterile option.
- 3-ply masks, individually bagged
- Sterile 10⁻⁶, gamma sterilization
- Duckbill with ties configuration
- Natural, latex-free
- Low particle shedding
VWR® Maximum Protection sterile masks provide superior particle and bacterial filtration efficiency plus low particle shedding. These moisture-resistant cleanroom masks consist of three layers featuring a unique, lightweight inner layer of film for enhanced resistance.
Suitable for use in cleanroom environments, production, and research.
Packaging: Masks are packaged 1 per bag, 20 bags per pouch bag, 10 pouch bags per case.