HALO® PAH, ENVIROCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
Certificates
About this item
HALO® ENVIROCLASS PAH columns are a performance-designed, application-specific phase to separate polycyclic aromatic hydrocarbons (PAH). HALO® ENVIROCLASS columns made with innovative Fused-Core® particle technology provide fast, high-resolution separations in a rugged column format designed for excellent reproducibility and column lifetimes. All ENVIROCLASS columns are lot tested and qualified to deliver an application-assured solution.
- Applicated solution for PAH analysis designed with environmental analysis needs in mind
- High-resolution and ultra-rugged performance
- Best Applications: EPA 610, EPA 8310, EU 15+1
- Optimal 2.7 µm Fused-Core® particle for rugged, reliable performance delivering high efficiency, low back pressure separations
- Pore size of these columns is proprietary
HALO® guard columns are available for this phase.
Specifications
- Catalog No:
- 76445-064
- 76445-060
- 76445-062
- 76445-074
- 76445-076
- 76445-066
- 76445-068
- 76443-516
- 76445-070
- 76443-514
- 76445-072
- 76443-512
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Temperature limit:
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)