SimpleDist® Filter Membranes, Environmental Express®
SimpleDist® micro replacement top membranes, Plastic
Catalog # 76436-868
Supplier: Environmental Express
Supplier #: C8250
SimpleDist® Filter Membranes, Environmental Express®
Catalog # 76436-868
Supplier: Environmental Express
Jump to:
Product Details & Documents
Prevent sample-to-sample contamination.
- Resistant to strong acids and bases
Ideal replacement membrane filters for your radiological extration filter funnels and SimpleDist® micro tubes.
Specifications
- Désignation:SimpleDist® micro replacement top membranes
- Matière:Plastic