StepSaver™ Individual Extraction Stations, Environmental Express®
StepSaver™ Individual Extraction Stations, Environmental Express®
StepSaver™ Individual Extraction Stations, Environmental Express®
Certificates
StepSaver™ Individual Extraction Stations, Environmental Express®
StepSaver™ Individual Extraction Stations, Environmental Express®
6 Options Available Below
Product Details & Documents
Constructed out of durable PVC and plastic materials, these compact, single-place extraction stations make oil and grease extractions easy.
  • Can be daisy-chained together to run multiple samples at one
  • Meets EPA method 1664 requirements for oil and grease extractions
  • Two separate chambers help keep water out of your extract
They are designed to be used for any type of Solid Phase Extraction (SPE) technique using an extraction disk. Single stations can be daisy-chained together to run multiple samples at once for faster sample throughput.
StepSaver™ system allows you to easily and efficiently collect the solvent with eluted oil and grease directly into a collection flask. No need to disassemble or reassemble glassware. As fluid enters the collection flask, it passes through a drying cartridge to remove excess water. At this point it is ready to be evaporated without a separate drying step.
The extraction stations are available in three different configurations: disposable filter funnel with UtraPrep® extraction disk, 47 mm glass filter funnel, and 90 mm glass filter funnel.
Product Family Options
Product Information
  • Désignation
    Includes
    Availability
    Price
  • StepSaver™ complete system, disposable 47 mm funnel
    Stepsaver base, inner sleeve, outer sleeves, funnel lid, hose barb, nylon plug, male luer sodium sulfate cartridge connector, fluorosilicone sealing gasket, funnel clamp, waste carboy with tubing and fittings, round bottom collection flask, funnels, aluminum weigh dishes, drying cartridge, O/G MDL standard with 7 mg HEM, and O/G standard with 40 mg HEM.
    Log in for availability
    Log in for availability
    Each
    Each - 3 522,37 $
    3 522,37 $
    Each
       3 522,37 $
    Minimum order quantity is 1
    Supplier Number: G6000
  • StepSaver™ individual station, disposable 47 mm funnel
    StepSaver base, inner sleeve, outer sleeves, funnel lid, hose barb, nylon plug, male luer sodium sulfate cartridge connector, fluorosilicone sealing gasket, funnel clamp, waste carboy with tubing and fittings, and round bottom collection flask.
    Log in for availability
    Log in for availability
    Each
    Each - 2 064,37 $
    2 064,37 $
    Each
       2 064,37 $
    Minimum order quantity is 1
    Supplier Number: G6001
  • StepSaver™ complete system, 47 mm glass funnel
    StepSaver base, inner sleeve, outer sleeves, glass filter funnel, SS screen for funnel base, funnel lid, hose barb, nylon plug, male luer sodium sulfate cartridge connector, fluorosilicone sealing gasket, funnel clamp, waste carboy with tubing and fittings, round bottom collection flask, ultraprep preactivated discs, aluminum weigh dishes, drying cartridge, O/G MDL standard with 7 mg HEM, and O/G standard with 40 mg HEM.
    Log in for availability
    Log in for availability
    Each
    Each - 3 883,41 $
    3 883,41 $
    Each
       3 883,41 $
    Minimum order quantity is 1
    Supplier Number: G6047
  • StepSaver™ individual station, 47 mm glass funnel
    StepSaver base, inner sleeve, outer sleeves, glass funnel, funnel lid, hose barb, nylon plug, male luer sodium sulfate cartridge connector, fluorosilicone sealing gasket, funnel clamp, waste carboy with tubing and fittings, and round bottom collection flask.
    Log in for availability
    Log in for availability
    Each
    Each - 2 429,38 $
    2 429,38 $
    Each
       2 429,38 $
    Minimum order quantity is 1
    Supplier Number: G6048
  • StepSaver™ complete system, 90 mm glass funnel
    StepSaver base, inner sleeve, outer sleeves, glass filter funnel, SS screen for funnel base, funnel lid, hose barb, nylon plug, male luer sodium sulfate cartridge connector, fluorosilicone sealing gasket, funnel clamp, waste carboy with tubing and fittings, round bottom collection flask, ultraprep preactivated discs, aluminum weigh dishes, drying cartridge, O/G MDL standard with 7 mg HEM, and O/G standard with 40 mg HEM.
    Log in for availability
    Log in for availability
    Each
    Each - 4 501,72 $
    4 501,72 $
    Each
       4 501,72 $
    Minimum order quantity is 1
    Supplier Number: G6090
  • StepSaver™ individual station, 90 mm glass funnel
    StepSaver base, inner sleeve, outer sleeves, glass funnel, funnel lid, hose barb, nylon plug, male luer sodium sulfate cartridge connector, fluorosilicone sealing gasket, funnel clamp, waste carboy with tubing and fittings, and round bottom collection flask.
    Log in for availability
    Log in for availability
    Each
    Each - 3 003,81 $
    3 003,81 $
    Each
       3 003,81 $
    Minimum order quantity is 1
    Supplier Number: G6091