HotBlock® 200 Digestion Controllers, Environmental Express®
Control one or two block heaters.
- Choose your controller, then block (or blocks) to suit your metals digestion needs
- Mount controllers outside the fume hood on metal surfaces, in an available outlet, or place on a benchtop
- Same dependability with increased temperature range
Perform a wider range of experiments and digestions with an expanded temperature range up to 200 °C with the new HotBlock 200. Controllers feature automatic shut-off, alarms, and ability to save profiles with ramp and soak settings. The dual-block controller can control up to two blocks independently and simultaneously. Safely place the external controller outside the fume hood. Controllers may be mounted to a metal surface using the magnets, or on an electrical outlet.
: Blocks and controllers are sold separately.
Frequently Bought Together