TKN System Controllers, Environmental Express® | Avantor
TKN System Controllers, Environmental Express®
TKN System Controllers, Environmental Express®
Certificates
TKN System Controllers, Environmental Express®
TKN System Controllers, Environmental Express®
2 Options Available Below
Product Details & Documents
Components to build your TKN system.
  • Controllers available as simple or advanced-touchscreen units
  • Up to 8 methods with 20 steps each can be stored in the touch screen controller
Temperature, ramp rate, and hold times are already programmed for the procedure. Works with the 24 and 54-well TKN HotBlocks, TKN028 and TKN054. The controller continually measures block temperature throughout the digestion process. All electrical components are housed in the controller away from the acid environment of your hood.
Product Family Options
Informations sur le produit
  • Désignation
    Control Type
    Availability
    Price
  • Environmental Express® HotBlock TKN100 simple system controller
    Digital
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,607.19
    $4,607.19
    Each
       $4,607.19
    La quantité minimale de commande est de 1
    Supplier Number: TKN100
    Spécifications:
    More
    Specifications
    Cat. No.
    76437-180
  • Environmental Express® HotBlock TKN180 touchscreen system controller
    Touchscreen digital
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,085.00
    $9,085.00
    Each
       $9,085.00
    La quantité minimale de commande est de 1
    Supplier Number: TKN180
    Spécifications:
    More
    Specifications
    Cat. No.
    76437-182