SimpleDist® Complete Distillation Systems, Environmental Express®
SimpleDist® Complete Distillation Systems, Environmental Express®
SimpleDist® Complete Distillation Systems, Environmental Express®
SimpleDist® Complete Distillation Systems, Environmental Express®
Safe and easy distillation system. The optimal system for performing cyanide, phenol, and ammonia distillations following established EPA methods 335.4 and 350.1, and Standard Methods 4500-CN-C and 4500-NH3.
  • Conform to EPA and Standard Methods
  • System reduces labor and clean-up time
  • Unique design replaces much of the fragile glassware usually involved in such procedures
The distillate is collected in a disposable trap, and does not require the use of a chiller or water lines. The sample boils in the glass boiling tube, and the gas is pulled by vacuum into the trap where it is collected. A hydrophobic porous frit in the trap prevents solution from flowing back into the boiling tube. When complete, the trap is removed and the contents are ready for analysis. System comes complete with everything for analysis.
 : Supplied with HEPA filter and an extra exhaust fan.
Product Family Options
  • Désignation
    Capacité
    Ø
    Temperature
    Shipping Weight
  • SimpleDist® complete distillation system, 120 VAC, max. 150 °C
    50 ml
    19.5×11.25×25"
    Ambient to 150 °C
    40 lbs.
    Each
    Each - $17,437.81
    $17,437.81
    Each
       $17,437.81
     : C6000
  • SimpleDist® complete distillation system, 240 VAC, max. 150 °C
    50 ml
    19.5×11.25×25"
    Ambient to 150 °C
    40 lbs.
    Each
    Each - $17,606.87
    $17,606.87
    Each
       $17,606.87
     : C6000-240
Accessories
  •  
    Description
  • 76436-836
    SimpleDist® two-port cap inserts, 38 mm, PP
    Pack of 6 - $218.46
  •  
    Description
  • 76436-834
    SimpleDist® washers for 38 mm cap
    Pack of 6 - $63.55
  •  
    Description
  • 76436-832
    SimpleDist® open top enclosure, 38 mm
    Pack of 6 - $311.09
  •  
    Description
  • 76436-826
    SimpleDist® 12-place tube transfer rack
    Each - $544.57
  •  
    Description
  • 76436-830
    SimpleDist® acid addition tube with funnel tip, PP
    Pack of 100 - $77.93
  •  
    Description
  • 76436-828
    SimpleDist® collection traps, 25 ml, pp
    Pack of 100 - $582.40
  •  
    Description
  • 76436-838
    SimpleDist® tubing adapters, pp
    Pack of 12 - $176.35
  •  
    Description
  • 76436-840
    SimpleDist® tubing kit, silicone and PP
    Pack of 12 - $209.53