Digestion Cups, Environmental Express®
Digestion Cups, Environmental Express®
Digestion Cups, Environmental Express®
Certificates
Digestion Cups, Environmental Express®
Digestion Cups, Environmental Express®
7 Options Available Below
Product Details & Documents
Disposable digestion cups eliminate transfer steps.
  • Cups that act as a digestion vessel or storage container and fit perfectly into the HotBlock and HotBlock Pro systems
  • Low metals background and EPA approved for digestions
Approved for hot plate digestions and certified for low metals background.
Documents
Product Family Options
Informations sur le produit
  • Capacité
    Matière
    Désignation
    Availability
    Price
  • 15 ml
    PP
    Digestion cups, natural screw-top caps
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1,000
    Pack of 1,000 - $382.44
    $382.44
    Pack of 1,000
       $382.44
    Supplier Number: SC415
  • 250 ml
    PP
    Digestion vessel with screw cap
    Discontinued
    Supplier Number: SC425
  • 100 ml
    PP
    Digestion cups, white screw-top caps
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 225
    Pack of 225 - $268.52
    $268.52
    Prix promotionnel pour les nouveaux clients
    Pack of 225
       $268.52
    Supplier Number: SC490
  • 50 ml
    PP
    UltimateCup, digestion cups with green caps
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 500
    Pack of 500 - $391.00
    $391.00
    Pack of 500
       $391.00
    Supplier Number: UC475-GN
  • 50 ml
    PP
    UltimateCup, digestion cups with natural linerless caps
    Discontinued
    Supplier Number: -
  • 50 ml
    PP
    UltimateCup, digestion cups with orange linerless cap
    Discontinued
    Supplier Number: UC475-OR
  • 50 ml
    PP
    UltimateCup, digestion cups with pink linerless cap
    Discontinued
    Supplier Number: UC475-PK
  • 50 ml
    PP
    UltimateCup, digestion cups with red linerless cap
    Discontinued
    Supplier Number: UC475-RD
  • 50 ml
    PP
    UltimateCup, digestion cups with white caps
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 500
    Pack of 500 - $420.39
    $420.39
    Pack of 500
       $420.39
    Supplier Number: UC475-WH
  • 50 ml
    PP
    UltimateCup, digestion cups with yellow linerless cap
    Discontinued
    Supplier Number: UC475-YL
  • 50 ml
    PP
    UltimateCup, digestion cups with natural linerless caps, rack lok
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 500
    Pack of 500 - $373.16
    $373.16
    Pack of 500
       $373.16
    Supplier Number: UC482-NL
  • 50 ml
    PP
    UltimateCup, digestion cups with white caps, rack lok
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 500
    Pack of 500 - $349.99
    $349.99
    Pack of 500
       $349.99
    Supplier Number: UC482-WH
  • 50 ml
    PP
    Ultimate clean cup, digestion cups with natural linerless caps
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 150
    Box of 150 - $343.86
    $343.86
    Box of 150
       $343.86
    Supplier Number: UCC000-50