ContactUsLinkComponent
EVA PAPR with 20LF2HE Loose-fitting Facepiece, Complete System, Bullard
EVA PAPR with 20LF2HE Loose-fitting Facepiece, Complete System, Bullard
Catalog # 76443-020
Supplier:  Bullard
undefined
EVA PAPR with 20LF2HE Loose-fitting Facepiece, Complete System, Bullard
Catalog # 76443-020
Supplier:  Bullard
Supplier Number:  EVAPAN4
Specifications
  • Description:
    EVA PAPR with 20LF2HE loose-fitting facepiece
  • Cat. no.:
    76443-020
Specifications
About this item
World Class PAPR that is ready to use directly out of the box.
  • All-in-one system ready for use, including one HEPA filter
  • Filters exhalated air to keep patients safe
  • Assigned protection factor (APF) 2.5X greater than an N95 mask
  • Improves breathing and overall comfort compared to an N95 mask
  • Loose-fitting facepiece designed for use with a stethoscope
  • No fit test
  • HEPA filter for use against communicable disease
EVA Powered Air-Purifying Respirator (PAPR) is equipped with the latest technology for providing wearers with constant airflow in a compact, streamlined, lightweight, ergonomic form for added comfort. Complete system includes a built-in HEPA exhalation panel to protect not only the PAPR wearer, but the environment and product as well.
Two selectable flow rate (low/high speeds) options: 7 cfm (200 lpm) and 8.5 cfm (240 lpm).
Certifications: NIOSH certified.
Ordering information: Complete EVA system includes EVA1 (blower unit), EVABELT1 (comfort closed cell foam belt), EVABAT1 (battery), EVASMC (single battery charger), PA1AFI (airflow indicator), PA20LFBT (breathing tube), 20LF2LHE facepiece, and HEPA filter.