UD 1 Series Safety Glasses, MCR Safety
Catalogue # 76343-354
Fournisseur: MCR Safety
Spécifications
- Frame Color:Gunmetal
- Description:Safety glasses VLT 90%
- Cat. no.:76343-354
- Lens Coating:Anti-fog (MAX6™)
- Lens Tint:Clear
- Frame Material:PC
Spécifications
À propos de cet article
UD1 Safety glasses are distinguished with application based, stylish design elements and superior coatings for work and play.
- Entire series meets military ballistic high velocity standard
- MAX6™ anti-fog coating is proven to provide six times greater fog-resistance
- Sleek co-injected TPR over polycarbonate temples
- Fixed closed cell foam lined options available
- Modern stylish design
The sleek and stylish modern design features non-slip temple grips, soft TPR nose pads, and crisp bayonet temples. These high-quality safety glasses have a lifetime frame warranty and are engineered to provide the highest level of innovation.
Certifications: ANSI Z87+. MIL - PRF - 31013.