To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Type:Dual swab
- Description:Transporter swab with amies gel, without charcoal
- Cat. no.:76447-102
- Packaging:50/box
- Tip Style:Rayon tip
- Application:Clinical
- Cap Color:Blue
Specifications
About this item
Transporter swab with amies gel, and without charcoal, features a dual plastic shaft, rayon swab tip, and blue cap.
- Unique tube shape preserves gel media
- Maximum gel for anaerobe preservation
- Available with or without charcoal
- Double plastic shaft
- Regular rayon tip
Amies media is Stuart's original media formula modified by replacing the glycerophosphate in the original formula for an inorganic phosphate buffer. The metabolism of glycerophosphate by coliform organisms and other Gram negative rods, in Stuart's original formulation, resulted in proliferation of these organisms from throat, wound, and fecal specimens. Amies also found that NaCl at 0.3% w/v was optimal for the preservation of Neisseria gonorrhoeae. Calcium and magnesium salts were added in the belief that these ions were of importance in controlling the permeability of the bacterial cells and contributed to their survival.