Monarch® RNA Cleanup Binding Buffer, New England Biolabs
CA76299-638
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Monarch® RNA cleanup binding buffer
- Size:80 ml
- Cat. no.:CA76299-638
This buffer is a component of the Monarch® RNA cleanup kits, is a guanidine-based buffer designed to dilute the sample and prepare the RNA sample to bind to the silica columns.
Monarch® kit components are available separately.