Monarch® RNA Cleanup Columns (500 µg), New England Biolabs
Catalog # CA76299-656
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:Monarch® RNA Cleanup Columns, 500 μg/100 columns
- Cat. no.:CA76299-656
Specifications
About this item
The Monarch® RNA cleanup columns (500 µg) are a component of the Monarch® RNA cleanup kit (500 µg) and can be used to purify up to 500 µg of RNA from enzymatic reactions including high-yield RNA synthesis reactions.
- Monarch® RNA Cleanup columns are also available for 10 and 50 µg binding capacities
The columns are designed without the use of a retaining ring, ensuring no buffer retention and no carryover of contaminants. RNA can be eluted in as little as 50 μl for and is ready for use in a variety of downstream applications including RT-PCR, RNA Library Prep for NGS and transfection.
Ordering information: RNA cleanup binding buffer and Monarch® RNA wash buffer are also available separately.
Delivery: Supplied with Monarch collection tubes II.