Monarch® RNA Cleanup Columns, New England Biolabs
Catalog # CA76307-362
Supplier: New England Biolabs (NEB)
CAS Number:
Specifications
- Description:Monarch® RNA Cleanup Columns, 10 μg/100 columns
- Cat. no.:CA76307-362
Specifications
About this item
The Monarch® RNA cleanup columns (10 µg) are a component of the Monarch RNA cleanup kit (10 µg) and can be used to purify up to 10 µg of RNA from enzymatic reactions. They are supplied with Monarch® collection tubes II. RNA cleanup binding buffer and Monarch® RNA wash buffer, are also available separately.
- Optimal for cleaning up RNA from a variety of enzymatic reactions
- Ideal for the concentration of RNA into small volumes (elute in ≥6 µl)
- 70 to 100% RNA recovery, even with inputs <20 ng
- Purified RNA is ready for use in a wide variety of downstream applications
- Unique column design prevents buffer carryover and elution of silica particulates
The columns are designed without the use of a retaining ring, ensuring no buffer retention and no carryover of contaminants. RNA can be eluted in as little as 6 μl and is ready for use in a variety of downstream applications including RT-PCR, RNA library prep for NGS, and labeling.
Monarch® RNA cleanup columns are also available for 50 and 500 µg binding capacities.