Temperature Probes, Environmental Express®
Temperature Probes, Environmental Express®
Certificates
Temperature Probes, Environmental Express®
Temperature Probes, Environmental Express®
3 Options Available Below
Product Details & Documents
Temperature probes enhance usage of Hot Blocks.
  • Compatible with Environmental Express® HotBlock® systems
The heads-up Lollipop™ Traceable® thermometer allows technicians to monitor actual sample temperature in 50 mL samples. The PTFE holder sits on the top of the digestion vessel. Thermometer has LCD display and can be set in Fahrenheit or Celsius. The stem is coated with PTFE to help in corrosive environments. Wide range covers 99 percent of the lab's routine temperature measurements. Solid-state construction eliminates dangers inherent in breakable glass and toxic mercury thermometers.
 : Kit contains the holder ring and thermometer with NIST Certification. NIST calibration good for at least 1 year from date of shipment.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Environmental Express® temperature probe, max. temp. 150 °C, for 15 ml digestion cup
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $436.78
    $436.78
    Each
       $436.78
    Supplier Number: SC985
  • Environmental Express® temperature probe, max. temp. 150 °C, for 50 ml digestion cup
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $446.41
    $446.41
    Each
       $446.41
    Supplier Number: SC980
  • Environmental Express® temperature probe, max. temp. 150 °C, for 100 ml digestion cup
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $436.78
    $436.78
    Each
       $436.78
    Supplier Number: SC981