Order Entry
Canada
ContactUsLinkComponent
Can-Terp Kit (High Level), 42 Components, SPEX CertiPrep
Can-Terp Kit (High Level), 42 Components, SPEX CertiPrep
Catalog # SXCANTERPKIT-H
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Can-Terp Kit (High Level), 42 Components, SPEX CertiPrep
Catalog # SXCANTERPKIT-H
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CAN-TERP-KIT-H
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml (1000 ppm)
  • Matrix:
    Methanol
  • Pack type:
    Ampoule, amber
  • Description:
    Can-Terp Kit (High Level), 42 components
  • Size:
    1 ml
  • Cat. no.:
    SXCANTERPKIT-H

Specifications

About this item

Can-Terp Mixes are analytical standards for medicinal and recreational cannabis testing.
Designed for Methods: State specific pesticide regulations:

  • OAR 333-008-11
  • HB 3460
  • AOAC 2007-01
  • EN 15662

While the legalization of cannabis for both medicinal and recreational purposes has been gaining speed, legislation and regulation has not necessarily kept pace. Out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are of high quality and free of chemical contaminants.

SPEX CertiPrep offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.