SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express® | Avantor
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®
Certificates
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®
SimpleWater™ BOD Nutrient Buffer Solutions, Environmental Express®
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
No mess - simply pour contents into your container and add water.
  • Solutions provide optimal growth media for bacteria in the BOD test
  • Borosilicate glass vials with leakproof screw caps
BOD SimpleWater™ Nutrient Buffer Dilution Vials are available in five standard concentrations. BOD dilution water is composed of a variety of nutrients that provide an optimal growth media for bacteria in the BOD test. SimpleWater BOD nutrient buffer solutions are concentrates of the four required nutrient solutions combined in one vial. The contents of the SimpleWater vial are then diluted to the appropriate volume to make up your BOD dilution water.
Packaging: Vials are borosilicate glass with lined, leak-proof screw caps. Glass vials are easy-to-rinse with no residue trapping.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • SimpleWater™ BOD Nutrient Buffer Solution
    6 L
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 36 (6l)
    Pack of 36 (6l) - $123.38
    $123.38
    Pack of 36 (6l)
       $123.38
    Supplier Number: D4006L
  • SimpleWater™ BOD Nutrient Buffer Solution
    19 L
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 36 (19l)
    Pack of 36 (19l) - $167.43
    $167.43
    Pack of 36 (19l)
       $167.43
    Supplier Number: D4019L
  • SimpleWater™ BOD Nutrient Buffer Solution
    55 L
    Discontinued
    Supplier Number: -
  • BOD: SimpleWater™ (Custom Size)
    9 L
    Discontinued
    Supplier Number: -
  • BOD: SimpleWater™ (Custom Size)
    10 L
    Discontinued
    Supplier Number: -
  • BOD: SimpleWater™ (Custom Size)
    16 L
    Discontinued
    Supplier Number: -
  • BOD: SimpleWater™ (Custom Size)
    24 L
    Discontinued
    Supplier Number: -
  • BOD: SimpleWater™ (Custom Size)
    31 L
    Discontinued
    Supplier Number: -