COD Reagent Vials, Environmental Express®
Obtain consistent and accurate results while saving money.
- Pre-filled reagent vials for either EPA-approved or mercury-free COD analysis
- Quality analyzed to provide you hassle-free consistent results
- Don''t change your instrument or method—vials work with Hach, Orbeco, Chemetrics, and Thermo COD systems
Pre-filled with COD reagents, these digestion vials come in either EPA-approved or mercury-free versions for low-, standard-, and ultra-high-range analysis. Each lot is tested to ensure superior quality from the cap and vial to the chemical contents. Glass vials are 16 mm x 100 mm to work with your existing COD reactor and spectrophotometer. Simply add 2 mL of your sample, heat in your digestor, and read in your spectrometer.
Although mercury-free vials are not approved for EPA Methods, they may be used for non-EPA COD testing when chlorine interferrence is not a factor. Ultra-high-range reagent vials have also not approved for EPA testing methods. COD digestion vials may contain acid, mercury, and silver and must be disposed of properly.
: Supplied with C of A and SDS.
Obtain consistent and accurate results while saving money.
- Pre-filled reagent vials for either EPA-approved or mercury-free COD analysis
- Quality analyzed to provide you hassle-free consistent results
- Don''t change your instrument or method—vials work with Hach, Orbeco, Chemetrics, and Thermo COD systems
Pre-filled with COD reagents, these digestion vials come in either EPA-approved or mercury-free versions for low-, standard-, and ultra-high-range analysis. Each lot is tested to ensure superior quality from the cap and vial to the chemical contents. Glass vials are 16 mm x 100 mm to work with your existing COD reactor and spectrophotometer. Simply add 2 mL of your sample, heat in your digestor, and read in your spectrometer.
Although mercury-free vials are not approved for EPA Methods, they may be used for non-EPA COD testing when chlorine interferrence is not a factor. Ultra-high-range reagent vials have also not approved for EPA testing methods. COD digestion vials may contain acid, mercury, and silver and must be disposed of properly.
: Supplied with C of A and SDS.
Product Family Options
- DésignationTailleRangeNo. of Tests
- COD reagent tubes Hg-freeVial 16 x 100 mm0 - 150 mg/L100/packPack of 1$375.45: B0010
- COD reagent tubes Hg-freeVial 16 x 100 mm0 - 1500 mg/L100/packPack of 1$375.45: B0015
- COD reagent tubes Hg-freeVial 16 x 100 mm0 - 15000 mg/L98/packPack of 98$386.23: B0020
- COD reagent tubes with HgVial 16 x 100 mm0 - 150 mg/L100/packPack of 1$375.33: B1010
- COD reagent tubes with HgVial 16 x 100 mm0 - 1500 mg/L100/packPack of 1$382.64: B1015
- COD reagent tubes with HgVial 16 x 100 mm0 - 15000 mg/L98/packPack of 98$375.45: B1020