Universal Solids Standard, Environmental Express®
Universal Solids Standard, Environmental Express®
Certificates
Universal Solids Standard, Environmental Express®
Universal Solids Standard, Environmental Express®
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Universal Solids Standard (USS) provides known QC data for all solids procedures.
  • Obtain batch level quality analysis and control for all solids testing at an affordable price
  • Quality guaranteed—NIST-traceability is shown on the Certificate of Analysis and each parameter has an acceptability range of ±3 standard deviations
  • Choose from three ranges to best fit your testing needs
This product complies with the following methods: SM 2540,ASTM D5907
These Environmental Express universal solids standards cover multiple EPA test method standard control requirements including TS, TSS, VS, and VSS. Use is easy—simply add the bottle contents into your graduated cylinder and fill to volume, or use as a Matrix Spike by adding a bottle directly to your sample.
 : Certificate of Analysis with traceability to NIST, SDS, and dilution instructions.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Type de conditionnement
    Availability
    Price
  • Universal Soilds Standard, Low Range
    10/pk
    Bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $175.34
    $175.34
    Pack of 10
       $175.34
    Supplier Number: F95005
  • Universal Soilds Standard, Ultra High Range
    10/pk
    Bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $231.35
    $231.35
    Pack of 10
       $231.35
    Supplier Number: F95005UH