COD Standards, Environmental Express®
COD Standards, Environmental Express®
COD Standards, Environmental Express®
COD Standards, Environmental Express®
Controls approved for use in both EPA-compliant and mercury-free vials.
  • Made from primary standard grade potassium hydrogen phthalate
  • Delivered with an SDS and minimum 8 month shelf life (refrigeration required)
  • Easy-to-follow dilution directions printed directly on the label
 : This product complies with the following methods: EPA 410.4, SM 5220D-2011, HACH 8000, ASTM D1252-06, ISO 15705:2002, USGS I-3561-06, BS 6068-2.34:1988, DIN 38414-9, and ISO 6060:1989.
Product Family Options
  • Désignation
    Taille
    Concentration
  • COD standard
    250 ml
    1000 mg/L
    Each (250ml)
    Each (250ml) - $73.54
    $73.54
    Each (250ml)
       $73.54
     : B1030
  • COD standard
    250 ml
    10000 mg/L
    Each (250ml)
    Each (250ml) - $73.54
    $73.54
    Each (250ml)
       $73.54
     : B1031