Crusher/Splitter Hoods
Supplier: MONASHEE MANUFACTURING CA
Certificates
About this item
The crusher/splitter hoods effectively capture and collect dust while providing uniform airflow, a clean work surface and protecting the operator from contaminants. Hoods can have the option to have your crusher placed on the left- or right-hand side to provide a convenient work area for a sample splitter. Designed for mining applications, our crusher/splitter hoods have a 10" flange for connecting to existing duct work and are required to be connected to an appropriately sized dust collector (sold separately, contact us for more information).
- Solid steel construction with powder coating for long lasting finish
- 10" flange to attach to ducting
- LED lighting
- Compressed air hose (air compressor sold separately)
- Crusher can be positioned on either the right or left-hand side
Crusher/splitter hoods are made of steel and blue powder coated for a long-lasting finish (additional colors available on request). Custom sizes are also available.
Frequently Bought Together