Certified pH Buffer Standards, SPEX CertiPrep
Certified pH Buffer Standards, SPEX CertiPrep
Certified pH Buffer Standards, SPEX CertiPrep
Supplier:  SPEX CERTIPREP LLC
Certificates
Certified pH Buffer Standards, SPEX CertiPrep
11 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Certified pH buffers are made using high purity starting materials and ASTM Type I Water, and are thoroughly analyzed for certified value and homogeneity. Buffers are accurate, premium grade for calibration and quality control checks.
  • Conveniently packaged in 500 ml LDPE bottles.
Certifications: SPEX CertiPrep pH Buffers are accurate and certified to ISO/IEC 17025:2005 and ISO 17034:2016 requirements.
Documents
Product Family Options
Product Information
  • Taille
    pH
    Color
    Availability
    Price
  • 500 ml
    2
    Colorless
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $117.83
    $117.83
    Each (500ml)
       $117.83
    Supplier Number: PH-BUFF2-500
  • 500 ml
    3
    Colorless
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $83.82
    $83.82
    Each (500ml)
       $83.82
    Supplier Number: PH-BUFF3-500
  • 500 ml
    4
    Red
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $109.96
    $109.96
    Each (500ml)
       $109.96
    Supplier Number: PH-BUFF4-500
  • 500 ml
    5
    Colorless
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $83.82
    $83.82
    Each (500ml)
       $83.82
    Supplier Number: PH-BUFF5-500
  • 500 ml
    6
    Colorless
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $117.83
    $117.83
    Each (500ml)
       $117.83
    Supplier Number: PH-BUFF6-500
  • 500 ml
    7
    Yellow
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $109.96
    $109.96
    Each (500ml)
       $109.96
    Supplier Number: PH-BUFF7-500
  • 500 ml
    8
    Colorless
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $109.96
    $109.96
    Each (500ml)
       $109.96
    Supplier Number: PH-BUFF8-500
  • 500 ml
    9
    Colorless
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $78.21
    $78.21
    Each (500ml)
       $78.21
    Supplier Number: PH-BUFF9-500
  • 500 ml
    10
    Blue
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $117.83
    $117.83
    Each (500ml)
       $117.83
    Supplier Number: PH-BUFF10-500
  • 500 ml
    11
    Colorless
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $135.80
    $135.80
    Each (500ml)
       $135.80
    Supplier Number: PH-BUFF11-500
  • 500 ml
    12
    Colorless
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $135.80
    $135.80
    Each (500ml)
       $135.80
    Supplier Number: PH-BUFF12-500