Ionic Strength Adjustment Buffers, SPEX CertiPrep
Buffers for Ion Chromarography and Ion Selective Electrode Standards
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. SPEX CertiPrep offers high quality IC standards available for the analytical laboratory.
Buffers for Ion Chromarography and Ion Selective Electrode Standards
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. SPEX CertiPrep offers high quality IC standards available for the analytical laboratory.
Product Family Options
- DésignationTailleType de conditionnement
- 5M Sodium nitrate buffer solution500 mlLDPE bottleEach (500ml)$307.62: IS-BUF1-500
- 10 M Sodium hydroxide buffer solution500 mlLDPE bottleEach (500ml)$339.59: IS-BUF2-500