Avantor® Hichrom HI-8 HT, GC Columns
Fournisseur: VWR International
Certificats
À propos de cet article
High Temperature general purpose column ideal for PCBs compounds, Pesticides, environmental analysis.
Maximum temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76431-292
- 76431-698
- 76431-696
- 76431-694
- 76431-692
- 76431-690
- 76431-688
- 76431-686
- 76431-684
- 76431-682
- 76431-680
- 76431-678
- 76431-676
- 76431-716
- 76431-714
- 76431-824
- 76431-712
- 76431-822
- 76431-710
- 76431-708
- 76431-706
- 76431-704
- 76431-702
- 76431-700
- Phase name:
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- HI-8 HT
- Phase composition:
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Proprietary Phase - high temperature
- Polarity:
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Low-to-mid polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C