Avantor® Hichrom HI-TCEP, GC Columns
Supplier: VWR International
Certificates
About this item
Application specifc columns ideal for aromatics and oxygenates in gasoline.
Maximum temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76429-106
- 76429-108
- 76429-102
- 76429-104
- Phase name:
- HI-TCEP
- HI-TCEP
- HI-TCEP
- HI-TCEP
- Phase composition:
- 1,2,3-tris-(2-cyanoethoxy)propane
- 1,2,3-tris-(2-cyanoethoxy)propane
- 1,2,3-tris-(2-cyanoethoxy)propane
- 1,2,3-tris-(2-cyanoethoxy)propane
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- Temperature limit:
- 150 °C
- 150 °C
- 150 °C
- 150 °C