Avantor® Hichrom HI-XMLB, GC Columns
Proprietary low-polarity and low-bleeding stationary phase; great choice for pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs). Ideal for GC/MS applications and is also an excellent choice as a confirmation column for semivolatile environmental analysis.
- Phase composition: crossbonded low-polarity proprietary phase
- Long lifetime
Max temperature limits may change depending on the film thickness.
- Catalog No:
- 76429-818
- 76429-836
- 76429-838
- 76429-832
- 76429-834
- 76429-828
- 76429-850
- 76429-830
- 76429-852
- 76429-824
- 76429-846
- 76429-826
- 76429-848
- 76429-820
- 76429-842
- 76429-822
- 76429-844
- 76429-840
- Phase name:
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- HI-XMLB
- Phase composition:
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Proprietary Phase developed for semi-volatiles environmental - low bleeding
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
Frequently Bought Together