Avantor® Hichrom HI-BASIC, GC Columns | Avantor
Avantor® Hichrom HI-BASIC, GC Columns
Supplier:   VWR International
Avantor® Hichrom HI-BASIC, GC Columns
Supplier:  VWR International
Certificates
Avantor® Hichrom HI-BASIC, GC Columns
2 Options Available Below
Product Details & Documents
Application specific column for basic compouns (e.g. amines).
Max temperature limits may change depending on the film thickness.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 76426-544
  • 76426-546
  • 76426-548
  • Phase name:
  • HI-BASIC
  • HI-BASIC
  • HI-BASIC
  • Phase composition:
  • Proprietary Phase developed for aliphatic and aromatics Amines
  • Proprietary Phase developed for aliphatic and aromatics Amines
  • Proprietary Phase developed for aliphatic and aromatics Amines
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Type de colonne
    Épaisseur du film
    Ø int.
    Longueur
    Availability
    Price
  • GC Column
    Capillaire
    0.15 µm
    0.25 mm
    25 m
    Discontinued
    Spécifications:
    More
    Specifications
    Cat. No.
    76426-544
    Phase name
    HI-BASIC
    Phase composition
    Proprietary Phase developed for aliphatic and aromatics Amines
  • GC Column
    Capillaire
    0.25 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,127.91
    $3,127.91
    Each
       $3,127.91
    Spécifications:
    More
    Specifications
    Cat. No.
    76426-546
    Phase name
    HI-BASIC
    Phase composition
    Proprietary Phase developed for aliphatic and aromatics Amines
  • GC Column
    Capillaire
    0.25 µm
    0.32 mm
    30 m
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,166.10
    $3,166.10
    Each
       $3,166.10
    Spécifications:
    More
    Specifications
    Cat. No.
    76426-548
    Phase name
    HI-BASIC
    Phase composition
    Proprietary Phase developed for aliphatic and aromatics Amines