Avantor® Hichrom HI-1701 HT, GC Columns
High temperature general purpose column. Ideal for confrmation analysis, ideal as mid-polar column in GC×GC-HT confgurations.
Max temperature limits may change depending on the film thickness.
- Catalog No:
- 76431-556
- 76431-578
- 76431-558
- 76431-552
- 76431-574
- 76431-554
- 76431-576
- 76431-548
- 76431-570
- 76431-550
- 76431-572
- 76431-812
- 76431-544
- 76431-566
- 76431-810
- 76431-546
- 76431-568
- 76431-540
- 76431-562
- 76431-542
- 76431-564
- 76431-560
- 76431-580
- Phase name:
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- HI-1701 HT
- Phase composition:
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- 14% Cyanopropylphenyl, 86% Methyl Polysiloxane - High Tempearature
- Polarity:
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Mid-polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- USP code:
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
- G46
Frequently Bought Together