Avantor® Hichrom HI-10, GC Columns
Fournisseur: VWR International
Certificats
À propos de cet article
Application specific highly polar phase, ideal for detailed cis/trans FAMEs separation.
- Phase composition: 100% cyanopropyl polysiloxane
Max temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76424-384
- 76424-382
- 76424-426
- 76424-386
- 76424-420
- 76424-418
- 76424-424
- 76424-422
- 76424-390
- 76424-434
- 76424-388
- 76424-432
- 76424-394
- 76424-438
- 76424-392
- 76424-436
- 76424-430
- 76424-428
- 76424-406
- 76424-404
- 76424-398
- 76424-396
- 76424-440
- 76424-402
- 76424-400
- 76424-412
- 76424-410
- 76424-416
- 76430-136
- 76424-414
- 76430-132
- 76430-134
- 76430-128
- 76424-408
- 76430-130
- Phase name:
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- HI-10
- Phase composition:
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- 100% Cyanopropyl Polysiloxane
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- Temperature limit:
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- 260 °C
- USP code:
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48
- G5, G8, G48