Avantor® Hichrom HI-DAI 1, GC Columns
Fournisseur: VWR International
Certificats
À propos de cet article
Application specifc column for direct introduction of aqueous samples, thus minimizing sample preparation.
Max temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76426-568
- 76426-572
- 76426-570
- 76426-566
- 76426-564
- Phase name:
- HI-DAI 1
- HI-DAI 1
- HI-DAI 1
- HI-DAI 1
- HI-DAI 1
- Phase composition:
- Proprietary Phase developed for direct aqueous injections
- Proprietary Phase developed for direct aqueous injections
- Proprietary Phase developed for direct aqueous injections
- Proprietary Phase developed for direct aqueous injections
- Proprietary Phase developed for direct aqueous injections
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C