Avantor® Hichrom HI-DAI 2, GC Columns
Application specifc column for direct introduction of aqueous samples, thus minimizing sample preparation.
Max temperature limits may change depending on the film thickness.
- Catalog No:
- 76426-584
- 76426-582
- 76426-576
- 76426-574
- 76426-580
- 76426-578
- Phase name:
- HI-DAI 2
- HI-DAI 2
- HI-DAI 2
- HI-DAI 2
- HI-DAI 2
- HI-DAI 2
- Phase composition:
- Proprietary phase developed for direct aqueous injections
- Proprietary phase developed for direct aqueous injections
- Proprietary phase developed for direct aqueous injections
- Proprietary phase developed for direct aqueous injections
- Proprietary phase developed for direct aqueous injections
- Proprietary phase developed for direct aqueous injections
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
- 320 °C
Frequently Bought Together