Avantor® Hichrom HI-PAH 2, GC Columns
Supplier: VWR International
Certificates
About this item
Application specifc column for polycyclic aromatics hydrocarbons.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76427-140
- 76427-136
- 76427-138
- 76427-134
- Phase name:
- HI-PAH 2
- HI-PAH 2
- HI-PAH 2
- HI-PAH 2
- Phase composition:
- Proprietary phase developed for EU PAHs analysis
- Proprietary phase developed for EU PAHs analysis
- Proprietary phase developed for EU PAHs analysis
- Proprietary phase developed for EU PAHs analysis
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 350 °C
- 350 °C
- 350 °C
- 350 °C