Avantor® Hichrom HI-17 HT, GC Columns
Supplier: VWR International
Certificates
About this item
High Temperature general purpose column. Ideal for confrmation analysis, ideal as high polarity dimension in GCxGC-HT confgurations.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76431-498
- 76431-538
- 76431-516
- 76431-518
- 76431-534
- 76431-512
- 76431-536
- 76431-514
- 76431-508
- 76431-530
- 76431-532
- 76431-510
- 76431-526
- 76431-504
- 76431-528
- 76431-506
- 76431-522
- 76431-500
- 76431-524
- 76431-502
- 76431-808
- 76431-806
- 76431-520
- Phase name:
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- Phase composition:
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- Polarity:
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Temperature limit:
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- USP code:
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17