Avantor® Hichrom HI-ALC 2, GC Columns
Fournisseur: VWR International
Certificats
À propos de cet article
Application specific column for blood alcohols testing.
Max temperature limits may change depending on the film thickness.
Spécifications
- Catalog No:
- 76426-540
- 76426-538
- 76426-542
- Phase name:
- HI-ALC 2
- HI-ALC 2
- HI-ALC 2
- Phase composition:
- Proprietary Phase developed for blood alcohol analysis
- Proprietary Phase developed for blood alcohol analysis
- Proprietary Phase developed for blood alcohol analysis
- Crossbond:
- Yes
- Yes
- Yes
- Temperature limit:
- 280 °C
- 280 °C
- 280 °C