Avantor® Hichrom HI-SE54 HT, GC Columns
Supplier: VWR International
Certificates
About this item
High Temperature general purpose column for high boiling petroleum products, Long-chained hydrocarbons.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76431-738
- 76431-736
- 76431-734
- 76431-732
- 76431-730
- 76431-728
- 76431-726
- 76431-724
- 76431-722
- 76431-720
- 76431-422
- 76431-718
- 76431-828
- 76431-826
- 76431-758
- 76431-756
- 76431-754
- 76431-752
- 76431-750
- 76431-748
- 76431-746
- 76431-744
- 76431-742
- 76431-740
- Phase name:
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- HI-SE54 HT
- Phase composition:
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- USP code:
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41