Avantor® Hichrom, Retention Gap/Precolumn, CW Deactivated Fused Silica
Supplier: VWR International
Certificates
About this item
CW deactivated fused silica retention gap / precolumn. Deactivation mainly for polar solvents.
Frequently Bought Together
Storage Rack for Vials, Polyethylene, Chemglass
Supplier: Chemglass
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
SureSTART™ Polyspring Inserts for 2 ml Vials, Level 2 High-Throughput, Thermo Scientific
Supplier: Thermo Scientific
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
SureSTART™ Caps for 2 ml Glass Snap Top Vials, Level 2 High-Throughput Applications, Thermo Scientific
Supplier: Thermo Scientific
Plastic Vials, Caps and Convenience Kits for LCMS and HPLC
Supplier: MicroSolv Technology Corp
Sample Containment Accessories, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
VWR® Ion Chromatography (IC) PolyVials
Supplier: VWR International
SureSTART™ Screw Caps for Glass Screw Top Vials, 9 mm, Level 1 Everyday Analysis, Thermo Scientific
Supplier: Thermo Scientific
Crimp Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Closures for 2 ml Crimp/Snap Top Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
SureSTART™ Crimp Caps, 11 mm, Level 3 High Performance Applications
Supplier: Thermo Scientific
WISP 48 Snap Seal Crimp-Top Vials, Restek
Supplier: Restek