AvantorĀ® Hichrom HI-VOC 2, GC Columns
Supplier: VWR International
Certificates
About this item
Application specifc columns for volatile organic compounds (VOCs), solvents and purgeable compounds. Due to the high maximum temperature, they are ideal for two-parallel columns confg. in the same oven. A good alternative to the HI-624 giving different selectivity. It is a more robust than the HI-624 and can operate at higher temperatures.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- 76429-196
- 76429-198
- 76429-276
- 76429-278
- 76429-272
- 76429-274
- 76429-268
- 76429-270
- 76429-264
- 76429-266
- 76429-282
- 76429-284
- 76429-280
- 76429-258
- 76429-254
- 76429-256
- 76429-250
- 76429-252
- 76429-246
- 76429-248
- 76429-242
- 76429-244
- 76429-260
- 76429-262
- 76429-236
- 76429-238
- 76429-232
- 76429-234
- 76429-228
- 76429-230
- 76429-224
- 76429-226
- 76429-220
- 76429-222
- 76429-240
- 76429-218
- 76429-214
- 76429-216
- 76429-210
- 76429-298
- 76429-212
- 76429-206
- 76429-294
- 76429-208
- 76429-296
- 76429-202
- 76429-290
- 76429-204
- 76429-292
- 76429-286
- 76429-200
- 76429-288
- Phase name:
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- HI-VOC 2
- Phase composition:
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Proprietary phase developed for volatile organic compounds
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes