HotBlock® COD Reactor, Environmental Express®
Supplier: Environmental Express
À propos de cet article
Digest two analytical batches at once!
- Process up to fifty-six standard, 16-mm diameter, round-bottom COD vials
- See actual and set-point temperatures simultaneously
- Pre-programmed to run at 150 °C for two hours, and flashes when the cycle is complete
- Maintain a setpoint of ±0.1 °C and sample consistency of ±1.5 °C
Reliable and uniform HotBlock technology now in a COD reactor. Process up to 56 samples, two analytical batches, in a single run! The digital display shows the actual temperature (in red) and set temperature (in green) simultaneously.
1 Options Available Below
Product Details & Documents
Digest two analytical batches at once!
- Process up to fifty-six standard, 16-mm diameter, round-bottom COD vials
- See actual and set-point temperatures simultaneously
- Pre-programmed to run at 150 °C for two hours, and flashes when the cycle is complete
- Maintain a setpoint of ±0.1 °C and sample consistency of ±1.5 °C
Reliable and uniform HotBlock technology now in a COD reactor. Process up to 56 samples, two analytical batches, in a single run! The digital display shows the actual temperature (in red) and set temperature (in green) simultaneously.
Specifications for Product Family
Spécifications
- Catalog No:
- 76436-812
- 76436-814
- Max. temperature:
- 150 °C
- 150 °C
- No. of tubes:
- 56 vials
- 56 vials
Spécifications
Product Family Options
240 V
Informations sur le produit
- DésignationElectricalAvailabilityPrice
- Supplier Number: B3000-240Spécifications:Irregular Voltage
More
Specifications
Cat. No.76436-814Max. temperature150 °CNo. of tubes56 vialsIrregular Voltage