Anti-POL II antibody will detect DNA-directed RNA polymerase II subunit RPB1, an enzyme composed of 12 subunits, which initiates the transcription of RNA from DNA, synthesizing many functional non-coding RNAs as well as messenger RNA precursors. POL II encodes a core element of the transcription machinery. Malfunctioning of the Pol II gene can cause diseases such as Cockayne Syndrome B and La Crosse Encephalitis. Anti-POL II Antibody is useful for researchers interested in DNA damage and repair studies.
Type: Primary
Antigen: POLR2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 76309-350
- 76310-450
- Antigen symbol:
- POLR2A
- POLR2A
- Antigen name:
- POL II
- POL II
- Antigen synonyms:
- RPO2, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, RNA-directed RNA polymerase II subunit RPB1, RPB1, POLR2A,
- RPO2, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, RNA-directed RNA polymerase II subunit RPB1, RPB1, POLR2A,
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P24928
- P24928
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid (sterile filtered)
- Liquid (sterile filtered)
- Purification:
- Affinity Purified
- Affinity Purified
- Storage buffer:
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Shipping temperature:
- Dry Ice
- Dry Ice