Virus Models Bundle
470329-776
: Avantor
- Description:Virus bundle
- Cat. no.:470329-776
Models and a kit.
- Mix of models and a kit
- Includes 10 'student' models
This bundle helps you to cover Viral structures. The HIV and Ebola models allow you to address specific viral pathogens, as well as general structure. Students can use the generic virion models to further explore virus structure, while assembling the vinyl models. Finally utilize the kit to have students build their own virus from scratch.
: Student models are intended for 1 to 2 assemblies. You may have different results, but repeated assembly and disassembly may cause breakage or excessive wear.