SP Wilmad-LabGlass Large Writing Area High-Throughput NMR Tubes, 5 mm O.D., SP Industries
Fournisseur: SP Industries
Certificats
À propos de cet article
Double the writing area allows for easy sample marking and identification.
- 2x larger marking area eliminates cramped and unreadable sample names
- Silkscreened writing area keeps sample names from easily wiping off
- Designed for routine use in most low to mid field NMR spectrometers
- Camber and concentricity values listed are the Total Indicator Reading (TIR)
- Made from ASTM E438 Type 1 Class B glass
- 100% inspected for surface defects and physical dimensions
These High-Throughput tubes have an average camber of 60 µm to ensure spectrum clarity for small molecule (MW<250) samples up to 600 MHz.
Frequently Bought Together