ContactUsLinkComponent
Magid® Clear Anti-Fog PET Full Face Shields with Soft Sponge Padding, Magid Glove
Catalog # 76417-632
Supplier:  Magid Glove
Magid® Clear Anti-Fog PET Full Face Shields with Soft Sponge Padding, Magid Glove
Catalog # 76417-632
Supplier:  Magid Glove
Supplier Number:  FS004
Specifications
  • Description:
    Clear anti-fog PET full face shield with soft sponge padding
  • Dimensions:
    8.6×13"
  • Cat. no.:
    76417-632
Specifications
About this item
Full faceshield features attached soft sponge padding at forehead, elastic band, and removable protective film.
  • Clear anti-fog PET full face shield protects the face from liquid splashes
  • Attached elastic band removes the need for bulky headgear
  • Comfortable soft sponge padding at forehead
  • Remove protective film before use
  • Caution – strap contains natural rubber latex, which may cause allergic reactions in some individuals
  • Not ANSI-rated for impact protection
Designed for use in applications requiring face protection from liquid splashes.
Certifications: Appropriate PPE face shield under OSHA 1910.1030 Occupational exposure to bloodborne pathogen and infectious materials.