SP Bel-Art Sterileware® Flat Bottom Sampling Scoops, White, Bel-Art Products, a part of SP
These sterile scoops are used by pharmaceutical, food, cosmetic and chemical companies for general production or laboratory work.
- Handle bump acts as thumb rest to help keep the hand away from the scoop; reduces contamination risks
- Flat bottom allows for use as a weigh boat
- Approximate volumes can be measured by removing excess powder
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10⁻⁶)
- Ready for one time use and disposal
- Lot stamped for accurate tracing; shipped with a certificate of processing for sterilization
These sampling scoops are designed for retrieving samples from a larger source. Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes.
: Sterilized products are not returnable.
Frequently Bought Together