ContactUsLinkComponent
QuantiChrom™ Hyaluronidase Inhibitor Screening Assay Kit, BioAssay Systems
  76423-336
 :  
undefined
QuantiChrom™ Hyaluronidase Inhibitor Screening Assay Kit, BioAssay Systems
  76423-336
 :  DIHY-100
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Hyaluronidase inhibitor screening assay kit
  • Cat. no.:
    76423-336
  • No. of Tests:
    100
High-throughput screening for evaluation of hyaluronidase inhibitors.
  • Rapid and reliable: entire assay can be completed in 30 minutes
  • Method: turbidimetric (OD 600 nm)
  • Samples: test compounds, hyaluronidase inhibitors
Hyaluronidases are a family of enzymes that catalyze the degradation of the glycosaminoglycan, hyaluronic acid. Hyaluronic acid is one of the major constituents of the extracellular matrix in organisms where it contributes to both cell proliferation and migration. The role of hyaluronidases in breaking down this key factor in cell proliferation makes them a possible target for cancer treatment. One hypothesis is that increased hyaluronidase may help prevent tumor invasion by breaking down the extracellular matrix needed for tumor expansion. Conversely, decreasing hyaluronidase activity might prevent metastasis by stopping cancer cells from escaping primary tumor masses. The study to determine hyaluronidases exact role in cancer pathology is still ongoing.
BioAssay Systems hyaluronidase inhibitor screening assay kit uses a two step turbidimetric reaction to measure hyaluronidase activity by the amount of hyaluronic acid that is hydrolyzed. A stop reagent halts the enzymatic reaction and forms turbidity with any residual hyaluronic acid in the well. The decrease in turbidity at 600 nm is directly proportional to hyaluronidase activity in the sample.