Corning® X‐LAB® Systems, Corning
About this item
The Corning® X‐LAB® systems are designed to separate and harvest a programmable volume of target cell populations from blood and blood products in a closed, sterile device.
- Density gradient media free sedimentation-based harvest
- Consistent, high recovery (>90%) of viable cells
- Process 40 to 240 ml
- Programmable harvest volume of 3 to 40 ml
- Barcode tracking of supplies and process steps electronically
- Single-use, disposable cartridge
Customizable process programming, automated separation of samples into three chambers, and highly sensitive infrared sensors for cell separation ensure consistent, reliable, efficient, high-recovery results.
Processing data can be transferred to X-SERIES® DataTrak software to further support MP processing, monitoring and reporting of information.
This system is a flexible solution for efficient, reliable, and reproducible density gradient media recovery of target cell populations from blood and blood products.
Certifications: US class I medical device.
4 Options Available Below
Product Details & Documents
The Corning® X‐LAB® systems are designed to separate and harvest a programmable volume of target cell populations from blood and blood products in a closed, sterile device.
- Density gradient media free sedimentation-based harvest
- Consistent, high recovery (>90%) of viable cells
- Process 40 to 240 ml
- Programmable harvest volume of 3 to 40 ml
- Barcode tracking of supplies and process steps electronically
- Single-use, disposable cartridge
Customizable process programming, automated separation of samples into three chambers, and highly sensitive infrared sensors for cell separation ensure consistent, reliable, efficient, high-recovery results.
Processing data can be transferred to X-SERIES® DataTrak software to further support MP processing, monitoring and reporting of information.
This system is a flexible solution for efficient, reliable, and reproducible density gradient media recovery of target cell populations from blood and blood products.
Certifications: US class I medical device.
Product Family Options
Product Information
- DésignationPackagingAvailabilityPrice
- Supplier Number: 6917
- Supplier Number: 6918
- Supplier Number: 6924
- Corning X‐LAB® disposable cartridges12/caseDiscontinued-Supplier Number: -
- Supplier Number: 6926
Accessories
Product Information
- Catalog #AvailabilityPrice
- Corning® X-Counterweight
Product Information
- Catalog #AvailabilityPrice
- Corning® X‐Balance Ring Kit, for balancing the contents of the centrifuge cup prior to centrifugationCorning® X‐Balance Ring Kit, for balancing the contents of the centrifuge cup prior to centrifugation76422-146Log in for availabilityCase of 1 - $533.00
Product Information
- Catalog #AvailabilityPrice
- Corning® X-SERIES® DataTrak software collects processing and system information through a user interface and stores it in a searchable, sortable databaseCorning® X-SERIES® DataTrak software collects processing and system information through a user interface and stores it in a searchable, sortable database76422-158Log in for availabilityCase of 1 - $2,140.00