À propos de cet article
PolarSafe® transportation kit together with a Traceable® data logger is the perfect combination to safely transport your critical samples while maintaining sample integrity. Choose between a PolarSafe® bag or box.
- Improve sample protection - Maintain specific temperature ranges for extended time periods
- Choose from durable transport bag or box to meet your needs
- Maintain sample integrity with Traceable® temperature monitoring
- Can be used as outer packaging according to P650 per UN3373 (category B)*
- Data logger provided with 2-year NIST-traceable calibration certificate
- Lowest set temperature of 2 °C
PolarSafe® Transport Bag Kits feature an insulated nylon bag and modular cooling packs for flexible configurations to fit your samples. Bag has pockets to keep documentation safe during transit, a shoulder strap, and carrying handles. The bundle adds a pack of 10 TraceableONE® single-use USB data loggers (model 18004-15). The logger is ready to use right out of the box – simply place it in your shipment and it will automatically generate a CSV report.
PolarSafe® Transport Box Kits feature polypropylene-insulated walls for improved insulation and modular cooling packs for flexible configurations to fit your samples. The box has both a shoulder strap and carrying handles for easy transportation. The bundle adds a pack of 10 TraceableONE™ single-use USB data loggers (model 18004-15). The logger is ready to use right out of the box – simply place it in your shipment and it will automatically generate a CSV report.